Anti-PSMC3 Mouse Monoclonal Antibody [clone: OTI7B10]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA500931EA 4340 DKK
ORIGTA500931
Anti-PSMC3 Mouse Monoclonal Antibody [clone: OTI7B10]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: PSMC3 (proteasome (prosome, macropain) 26S subunit, ATPase, 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR