Anti-ACO2 Mouse Monoclonal Antibody [clone: OTI3G10]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGTA500913EA 4500 DKK
ORIGTA500913
Anti-ACO2 Mouse Monoclonal Antibody [clone: OTI3G10]
Antistoffer
Recommended Dilutions: Immunofluorescence: 1:100 Flow Cytometry: 1:100
Type: Primary
Antigen: ACO2 (aconitase 2, mitochondrial)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human