Anti-ID3 Mouse Monoclonal Antibody [clone: OTI10D3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA500791EA 4500 DKK
ORIGTA500791
Anti-ID3 Mouse Monoclonal Antibody [clone: OTI10D3]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: ID3 (inhibitor of DNA binding 3, dominant negative helix-loop-helix protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR