Anti-SORD Mouse Monoclonal Antibody [clone: OTI13G5]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGTA500784EA 4500 DKK
ORIGTA500784
Anti-SORD Mouse Monoclonal Antibody [clone: OTI13G5]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50
Type: Primary
Antigen: SORD (sorbitol dehydrogenase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human