Anti-RNF144B Mouse Monoclonal Antibody [clone: OTI3C2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA500696EA 4500 DKK
ORIGTA500696
Anti-RNF144B Mouse Monoclonal Antibody [clone: OTI3C2]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: RNF144B (ring finger protein 144B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR