Anti-GBP2 Mouse Monoclonal Antibody [clone: OTI4E7]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA500660EA 4340 DKK
ORIGTA500660
Anti-GBP2 Mouse Monoclonal Antibody [clone: OTI4E7]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GBP2 (guanylate binding protein 2, interferon-inducible)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR