Anti-PPP1R7 Mouse Monoclonal Antibody [clone: OTI4F9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA500614EA 4380 DKK
ORIGTA500614
Anti-PPP1R7 Mouse Monoclonal Antibody [clone: OTI4F9]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100

Type: Primary
Antigen: PPP1R7 (protein phosphatase 1, regulatory subunit 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR