Anti-SATB1 Mouse Monoclonal Antibody [clone: OTI4E1]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA500524EA 4600 DKK
ORIGTA500524
Anti-SATB1 Mouse Monoclonal Antibody [clone: OTI4E1]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Immunofluorescence: 1:100

Type: Primary
Antigen: SATB1 (SATB homeobox 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR