Anti-SCYL3 Mouse Monoclonal Antibody [clone: OTI4F10]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA500493EA 4640 DKK
ORIGTA500493
Anti-SCYL3 Mouse Monoclonal Antibody [clone: OTI4F10]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Immunofluorescence: 1:100 Flow Cytometry: 1:1000

Type: Primary
Antigen: SCYL3 (SCY1-like 3 (S. cerevisiae))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR