Anti-IRF3 Mouse Monoclonal Antibody [clone: OTI5D2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA500465EA 4430 DKK
ORIGTA500465
Anti-IRF3 Mouse Monoclonal Antibody [clone: OTI5D2]
Antistoffer
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: IRF3 (interferon regulatory factor 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR