Anti-NRBP1 Mouse Monoclonal Antibody [clone: OTI6C11]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA500443EA 4600 DKK
ORIGTA500443
Anti-NRBP1 Mouse Monoclonal Antibody [clone: OTI6C11]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:50; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: NRBP1 (nuclear receptor binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR