Anti-SOX17 Mouse Monoclonal Antibody [clone: OTI5G6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA500282EA 4430 DKK
ORIGTA500282
Anti-SOX17 Mouse Monoclonal Antibody [clone: OTI5G6]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SOX17 (SRY (sex determining region Y)-box 17)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR