Anti-ACTN1 Mouse Monoclonal Antibody [clone: OTI7A4]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA500072EA 4380 DKK
ORIGTA500072
Anti-ACTN1 Mouse Monoclonal Antibody [clone: OTI7A4]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: ACTN1 (actinin, alpha 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR