Anti-BMP4 Mouse Monoclonal Antibody [clone: OTI6B7]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA500014EA 4500 DKK
ORIGTA500014
Anti-BMP4 Mouse Monoclonal Antibody [clone: OTI6B7]
Antistoffer
Recommended Dilutions: Western Blot: 1:100000; Immunofluorescence: 1:100

Type: Primary
Antigen: BMP4 (bone morphogenetic protein 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR