Anti-FGF2 Mouse Monoclonal Antibody [clone: OTI3D9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA500012EA 4650 DKK
ORIGTA500012
Anti-FGF2 Mouse Monoclonal Antibody [clone: OTI3D9]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: FGF2 (fibroblast growth factor 2 (basic))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR