Anti-ENTPD2 Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA318852EA 4970 DKK
ORIGTA318852
Anti-ENTPD2 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 15-20 µg/ml

Type: Primary
Antigen: ENTPD2 (ectonucleoside triphosphate diphosphohydrolase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR