Anti-IL10 Rat Polyclonal Antibody [clone: JES5-2A5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA318463EA 4970 DKK
ORIGTA318463
Anti-IL10 Rat Polyclonal Antibody [clone: JES5-2A5]
Antistoffer
Recommended Dilutions: ELISA Flo: 1:10-1:100; Immunochemistry: 10 µg/ml; Western Blot: 1:1000

Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR