Anti-MCAM Mouse Polyclonal Antibody [clone: P1H12]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA318345EA 5080 DKK
ORIGTA318345
Anti-MCAM Mouse Polyclonal Antibody [clone: P1H12]
Antistoffer
Recommended Dilutions: Flo; Immunochemistry: 20 µg/ml; Immunoprecipitation; Western Blot: 1-5 µg/ml

Type: Primary
Antigen: MCAM (melanoma cell adhesion molecule)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR