Anti-ROR1 Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA318277EA 4970 DKK
ORIGTA318277
Anti-ROR1 Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 3-6 µg/ml

Type: Primary
Antigen: ROR1 (receptor tyrosine kinase-like orphan receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR