Anti-MBP Mouse Monoclonal Antibody [clone: V/h5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA318238EA 5080 DKK
ORIGTA318238
Anti-MBP Mouse Monoclonal Antibody [clone: V/h5]
Antistoffer
  • Recommended Dilutions: Immunochemistry: 10 µg/ml

Type: Primary
Antigen: MBP (myelin basic protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR