Anti-MAOB Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA318153EA 4970 DKK
ORIGTA318153
Anti-MAOB Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: ELISA: 1:32000; Immunochemistry: 5 µg/ml; Western Blot: 1-3 µg/ml

Type: Primary
Antigen: MAOB (monoamine oxidase B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR