Anti-NLRP2 Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA317770EA 5920 DKK
ORIGTA317770
Anti-NLRP2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NLRP2 (NLR family, pyrin domain containing 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR