Anti-WNT7A Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA317522EA 5080 DKK
ORIGTA317522
Anti-WNT7A Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 20 µg/ml

Type: Primary
Antigen: WNT7A (wingless-type MMTV integration site family, member 7A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR