Anti-TNFRSF11A Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA317443EA 5080 DKK
ORIGTA317443
Anti-TNFRSF11A Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 10 µg/ml; Western Blot: 1-3 µg/ml

Type: Primary
Antigen: TNFRSF11A (tumor necrosis factor receptor superfamily, member 11a, NFKB activator)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR