Anti-TLL2 Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA317423EA 4970 DKK
ORIGTA317423
Anti-TLL2 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 20 µg/ml

Type: Primary
Antigen: TLL2 (tolloid-like 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR