Anti-SSTR5 Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA317368EA 5080 DKK
ORIGTA317368
Anti-SSTR5 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 23 µg/ml

Type: Primary
Antigen: SSTR5 (somatostatin receptor 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR