Anti-HPN Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA316896EA 4970 DKK
ORIGTA316896
Anti-HPN Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 5 µg/ml; Western Blot: 1 µg/ml

Type: Primary
Antigen: HPN (hepsin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR