Anti-DRD5 Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA316684EA 4970 DKK
ORIGTA316684
Anti-DRD5 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 20 µg/ml

Type: Primary
Antigen: DRD5 (dopamine receptor D5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR