Anti-CALCRL Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA316587EA 4970 DKK
ORIGTA316587
Anti-CALCRL Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 16 µg/ml

Type: Primary
Antigen: CALCRL (calcitonin receptor-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR