Anti-HIRA Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA314655EA 4120 DKK
ORIGTA314655
Anti-HIRA Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:1000

Type: Primary
Antigen: HIRA (histone cell cycle regulator)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR