Anti-ST6GAL1 Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA314536EA 4120 DKK
ORIGTA314536
Anti-ST6GAL1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: ST6GAL1 (ST6 beta-galactosamide alpha-2,6-sialyltranferase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR