Anti-RPS4Y1 Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA314477EA 4120 DKK
ORIGTA314477
Anti-RPS4Y1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: RPS4Y1 (ribosomal protein S4, Y-linked 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR