Anti-CACNG5 Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA308503EA 4560 DKK
ORIGTA308503
Anti-CACNG5 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot:1:500-1:3000

Type: Primary
Antigen: CACNG5 (calcium channel, voltage-dependent, gamma subunit 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR