Anti-THOC1 Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA308394EA 4560 DKK
ORIGTA308394
Anti-THOC1 Rabbit Polyclonal Antibody
Antistoffer
  • Recommended Dilutions:

Type: Primary
Antigen: THOC1 (THO complex 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR