Anti-LAMTOR3 Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA308268EA 4560 DKK
ORIGTA308268
Anti-LAMTOR3 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot:1:500-1:3000

Type: Primary
Antigen: LAMTOR3 (late endosomal/lysosomal adaptor, MAPK and MTOR activator 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR