Anti-POMGNT1 Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA308113EA 4060 DKK
ORIGTA308113
Anti-POMGNT1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: POMGNT1 (protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR