Anti-RPS4X Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA308015EA 4060 DKK
ORIGTA308015
Anti-RPS4X Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot:1:500-1:3000

Type: Primary
Antigen: RPS4X (ribosomal protein S4, X-linked)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR