Anti-UAP1 Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA307963EA 4560 DKK
ORIGTA307963
Anti-UAP1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: UAP1 (UDP-N-acteylglucosamine pyrophosphorylase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR