Anti-MYC Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA307883EA 4560 DKK
ORIGTA307883
Anti-MYC Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: FACS:1:50-1:200; Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR