Anti-RAPGEF3 Rabbit Monoclonal Antibody [clone: EPR1672]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA307796EA 5590 DKK
ORIGTA307796
Anti-RAPGEF3 Rabbit Monoclonal Antibody [clone: EPR1672]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunoprecipitation: 1:10-1:100; Immunochemistry: 1:50-1:100;

Type: Primary
Antigen: RAPGEF3 (Rap guanine nucleotide exchange factor (GEF) 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR