Anti-APOL1 Rabbit Monoclonal Antibody [clone: EPR2907(2)]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA307579EA 5120 DKK
ORIGTA307579
Anti-APOL1 Rabbit Monoclonal Antibody [clone: EPR2907(2)]
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunoprecipitation: 1:10-1:100; Immunochemistry: 1:100-1:250; Flow Cytometry: 1:10-1:100

Type: Primary
Antigen: APOL1 (apolipoprotein L, 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR