Anti-FADD Rabbit Monoclonal Antibody [clone: EPR4415]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA307340EA 5590 DKK
ORIGTA307340
Anti-FADD Rabbit Monoclonal Antibody [clone: EPR4415]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunoprecipitation: 1:10-1:100; Immunochemistry: 1:100-1:250

Type: Primary
Antigen: FADD (Fas (TNFRSF6)-associated via death domain)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR