Anti-CEBPB Rabbit monoclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA307242EA 3550 DKK
ORIGTA307242
Anti-CEBPB Rabbit monoclonal antibody
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry/Immunofluorescence: 1:50-1:100; Flow Cytometry: 1:1000; Immunoprecipitation: 1:20

Type: Primary
Antigen: CEBPB (CCAAT/enhancer binding protein (C/EBP), beta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR