Anti-CABS1 Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA306879EA 4180 DKK
ORIGTA306879
Anti-CABS1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CABS1 (calcium-binding protein, spermatid-specific 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR