Anti-SRPX Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA306869EA 4180 DKK
ORIGTA306869
Anti-SRPX Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.25 ?g/ml; Immunochemistry: 5 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: SRPX (sushi-repeat containing protein, X-linked)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR