Anti-VPS53 Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA306570EA 4480 DKK
ORIGTA306570
Anti-VPS53 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 5 ?g/ml; Immunofluorescence: 2.5 ?g/ml

Type: Primary
Antigen: VPS53 (vacuolar protein sorting 53 homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR