Anti-TNFRSF14 Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA306306EA 4180 DKK
ORIGTA306306
Anti-TNFRSF14 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 2 ?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: TNFRSF14 (tumor necrosis factor receptor superfamily, member 14)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR