Anti-CBLN1 Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA306244EA 4180 DKK
ORIGTA306244
Anti-CBLN1 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 2-4 ?g/ml

Type: Primary
Antigen: CBLN1 (cerebellin 1 precursor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR