Anti-DAGLA Goat Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA305875EA 3810 DKK
ORIGTA305875
Anti-DAGLA Goat Antibody
Antistoffer
Recommended Dilutions: Western Blot: 005-02?g/ml

Type: Primary
Antigen: DAGLA (diacylglycerol lipase, alpha)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR