Anti-GPR125 Goat Antibody
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGTA305855EA 4090 DKK
ORIGTA305855
Anti-GPR125 Goat Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1-3?g/ml
Type: Primary
Antigen: GPR125 (G protein-coupled receptor 125)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human